siRNA Details
| Gene Symbol: Acacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053922.1 | NP_446374.1 | 4 | 963 | ||
| XM_008769309.1 | XP_008767531.1 | 5 | 1446 | |||
| XM_008769310.1 | XP_008767532.1 | 5 | 2050 | |||
| XM_008769311.1 | XP_008767533.1 | 6 | 1555 | |||
| XM_008769312.1 | XP_008767534.1 | 5 | 1541 | |||
| XM_008769313.1 | XP_008767535.1 | 5 | 1446 | |||
| XM_008769314.1 | XP_008767536.1 | 4 | 801 | |||
| GenBank mRNA | AB004329.1 | - | 4 | 963 | ||
| DQ493871.1 | - | 4 | 986 | |||

Acacb
Genomic Map