siRNA Details
| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053949.1 | NP_446401.1 | 4 | 650 | ||
| XM_006235867.2 | XP_006235929.1 | 4 | 783 | |||
| GenBank mRNA | Z96106.1 | - | 4 | 650 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnh2| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053949.1 | NP_446401.1 | 4 | 650 | ||
| XM_006235867.2 | XP_006235929.1 | 4 | 783 | |||
| GenBank mRNA | Z96106.1 | - | 4 | 650 | ||