siRNA Details
| Gene Symbol: BRINP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014618.2 | NP_055433.2 | 2 | 572 | ||
| GenBank mRNA | AF027734.1 | - | 2 | 534 | ||
| AK293649.1 | - | 2 | 546 | |||
| BC021560.1 | - | 2 | 536 | |||
| BC065196.1 | - | 2 | 525 | 5245480 | ||

BRINP1
Genomic Map