siRNA Details
| Gene Symbol: Nln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053970.2 | NP_446422.2 | 2 | 127 | ||
| XM_006231880.2 | XP_006231942.1 | 2 | 281 | |||
| XM_006231881.2 | XP_006231943.1 | 3 | 359 | |||
| GenBank mRNA | BC072687.1 | - | 2 | 127 | 7099208 | |
| X87157.1 | - | 2 | 235 | |||

Nln
Genomic Map