siRNA Details
| Gene Symbol: RragB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053972.2 | NP_446424.1 | 2, 3 | 654 | ||
| XM_006256779.2 | XP_006256841.1 | 2, 3 | 752 | |||
| XM_008773066.1 | XP_008771288.1 | 1, 2 | 809 | |||
| XM_008773067.1 | XP_008771289.1 | 1, 2 | 809 | |||
| GenBank mRNA | BC078760.1 | - | 2, 3 | 192 | 7123902 | |
| BC087698.1 | - | 2, 3 | 654 | 7314028 | ||
| Gene Symbol: LOC100909655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752022.3 | XP_003752070.1 | 753 | |||
| XM_006256775.2 | XP_006256837.1 | 752 | ||||
| XM_008773107.1 | XP_008771329.1 | 809 | ||||
| XM_008773108.1 | XP_008771330.1 | 809 | ||||
| XM_008773109.1 | XP_008771331.1 | 1, 2 | 809 | |||

RragB
Genomic Map