siRNA Details
| Gene Symbol: Eif4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053974.2 | NP_446426.1 | 2 | 99 | ||
| XM_008761488.1 | XP_008759710.1 | 3 | 212 | |||
| GenBank mRNA | BC087001.1 | - | 2 | 99 | 7122606 | |
| FQ218941.1 | - | 2 | 91 | |||
| X83399.1 | - | 2 | 134 | |||

Eif4e
Genomic Map