siRNA Details
| Gene Symbol: Gjb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053984.2 | NP_446436.1 | 1 | 1093 | ||
| GenBank mRNA | X76168.1 | - | 1 | 1093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gjb4| Gene Symbol: Gjb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053984.2 | NP_446436.1 | 1 | 1093 | ||
| GenBank mRNA | X76168.1 | - | 1 | 1093 | ||