siRNA Details
| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053990.1 | NP_446442.1 | 3 | 266 | ||
| XM_006254854.2 | XP_006254916.1 | 3 | 1505 | |||
| XM_006254855.2 | XP_006254917.1 | 3 | 1505 | |||
| XM_008772080.1 | XP_008770302.1 | 3 | 1505 | |||
| XM_008772081.1 | XP_008770303.1 | 3 | 1505 | |||
| GenBank mRNA | BC078758.1 | - | 3 | 305 | 7132485 | |
| X58828.1 | - | 3 | 266 | |||

Ptpn2
Genomic Map