siRNA Details
| Gene Symbol: Ppp2r2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053999.2 | NP_446451.1 | 3 | 447 | ||
| XM_008770730.1 | XP_008768952.1 | 3 | 1254 | |||
| XM_008770731.1 | XP_008768953.1 | 3 | 1254 | |||
| GenBank mRNA | BC128731.1 | - | 3 | 447 | 7370343 | |
| M83297.1 | - | 3 | 342 | |||
| M83298.1 | - | 3 | 428 | |||

Ppp2r2a
Genomic Map