siRNA Details
| Gene Symbol: Acsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057107.1 | NP_476448.1 | 1 | 262 | ||
| GenBank mRNA | D30666.1 | - | 1 | 262 | ||
| FQ221868.1 | - | 472 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acsl3| Gene Symbol: Acsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057107.1 | NP_476448.1 | 1 | 262 | ||
| GenBank mRNA | D30666.1 | - | 1 | 262 | ||
| FQ221868.1 | - | 472 | ||||