siRNA Details
| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053958.1 | NP_446410.1 | 3 | 895 | ||
| XM_006244178.2 | XP_006244240.1 | 3 | 1673 | |||
| GenBank mRNA | Y13400.1 | - | 3 | 895 | ||
More Information
Other Species Targeted
Set Membership: |
|

Ccr3
Genomic Map