siRNA Details
| Gene Symbol: Gps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270939.1 | NP_001257868.1 | 3 | 284 | ||
| NM_053969.3 | NP_446421.3 | 3 | 458 | |||
| XM_006247865.1 | XP_006247927.1 | 3 | 453 | |||
| XM_006247866.1 | XP_006247928.1 | 3 | 453 | |||
| XM_006247867.2 | XP_006247929.1 | 3 | 313 | |||
| XM_008768462.1 | XP_008766684.1 | 3 | 453 | |||
| XM_008768463.1 | XP_008766685.1 | 3 | 313 | |||
| GenBank mRNA | BC061746.1 | - | 3 | 407 | 5599712 | |
| CB792560.1 | - | 3 | 335 | |||
| X87885.1 | - | 2 | 239 | |||

Gps1
Genomic Map