siRNA Details
Gene Symbol: Calb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053988.1 | NP_446440.1 | 2 | 212 | ||
GenBank mRNA | X66974.1 | - | 2 | 212 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Calb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053988.1 | NP_446440.1 | 2 | 212 | ||
GenBank mRNA | X66974.1 | - | 2 | 212 |