siRNA Details
| Gene Symbol: Kcnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054000.2 | NP_446452.2 | 2 | 766 | ||
| GenBank mRNA | M77482.1 | - | 2 | 769 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnb2| Gene Symbol: Kcnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054000.2 | NP_446452.2 | 2 | 766 | ||
| GenBank mRNA | M77482.1 | - | 2 | 769 | ||