siRNA Details
| Gene Symbol: Tcea2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057098.1 | NP_476439.1 | 8 | 882 | ||
| XM_006235740.1 | XP_006235802.1 | 8 | 974 | |||
| XM_006235741.1 | XP_006235803.1 | 8 | 959 | |||
| XM_006235742.2 | XP_006235804.1 | 8 | 948 | |||
| GenBank mRNA | D12927.1 | - | 8 | 882 | ||
| FQ213752.1 | - | 8 | 860 | |||

Tcea2
Genomic Map