siRNA Details
| Gene Symbol: Gas6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057100.2 | NP_476441.2 | 6 | 636 | ||
| GenBank mRNA | BC070881.1 | - | 6 | 636 | 7096665 | |
| D42148.1 | - | 6 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gas6| Gene Symbol: Gas6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057100.2 | NP_476441.2 | 6 | 636 | ||
| GenBank mRNA | BC070881.1 | - | 6 | 636 | 7096665 | |
| D42148.1 | - | 6 | 531 | |||