siRNA Details
| Gene Symbol: Tra2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057119.1 | NP_476460.1 | 3 | 340 | ||
| XM_006248536.2 | XP_006248598.1 | 3 | 381 | |||
| XM_008768789.1 | XP_008767011.1 | 3 | 4496 | |||
| XM_008768790.1 | XP_008767012.1 | 4 | 657 | |||
| GenBank mRNA | BC070948.1 | - | 3 | 340 | 7097246 | |
| D49708.1 | - | 3 | 340 | |||
| FQ212414.1 | - | 3 | 354 | |||

Tra2b
Genomic Map