siRNA Details
| Gene Symbol: Slc15a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057121.1 | NP_476462.1 | 6, 7 | 518 | ||
| XM_006252470.2 | XP_006252532.1 | 6, 7 | 675 | |||
| GenBank mRNA | D50306.1 | - | 6, 7 | 516 | ||
| D50664.1 | - | 6, 7 | 518 | |||

Slc15a1
Genomic Map