siRNA Details
| Gene Symbol: Psmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057122.1 | NP_476463.1 | 3 | 206 | ||
| GenBank mRNA | BC063145.1 | - | 3 | 216 | 6889167 | |
| D50695.1 | - | 3 | 206 | |||
| FQ225557.1 | - | 3 | 231 | |||
| FQ226113.1 | - | 3 | 215 | |||

Psmc4
Genomic Map