siRNA Details
| Gene Symbol: Nup155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053952.1 | NP_446404.1 | 11, 12 | 1326 | ||
| GenBank mRNA | Z21780.1 | - | 11, 12 | 1326 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup155| Gene Symbol: Nup155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053952.1 | NP_446404.1 | 11, 12 | 1326 | ||
| GenBank mRNA | Z21780.1 | - | 11, 12 | 1326 | ||