siRNA Details
| Gene Symbol: Sds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053962.3 | NP_446414.3 | 6 | 502 | ||
| XM_008769213.1 | XP_008767435.1 | 5 | 1000 | |||
| GenBank mRNA | BC088110.1 | - | 6 | 502 | 7323391 | |
| J03863.1 | - | 6 | 700 | |||
| Y00752.1 | - | 6 | 519 | |||

Sds
Genomic Map