siRNA Details
| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057207.3 | NP_476555.1 | 2 | 559 | ||
| GenBank mRNA | L10362.1 | - | 2 | 559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sv2b| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057207.3 | NP_476555.1 | 2 | 559 | ||
| GenBank mRNA | L10362.1 | - | 2 | 559 | ||