siRNA Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_058209.2 | NP_478116.2 | 173 | |||
| XM_008763803.1 | XP_008762025.1 | 2 | 470 | |||
| XM_008763804.1 | XP_008762026.1 | 2 | 467 | |||
| GenBank mRNA | AF072439.1 | - | 198 | |||
| BC166844.1 | - | 110 | ||||
| CB727538.1 | - | 2 | 444 | |||

Zfp37
Genomic Map