siRNA Details
| Gene Symbol: Apbb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080478.1 | NP_536726.1 | 1 | 152 | ||
| XM_006229935.1 | XP_006229997.1 | 2 | 521 | |||
| XM_006229936.2 | XP_006229998.1 | 2 | 357 | |||
| XM_006229938.2 | XP_006230000.1 | 2 | 696 | |||
| GenBank mRNA | AF333983.1 | - | 1 | 152 | ||

Apbb1
Genomic Map