siRNA Details
| Gene Symbol: Dgka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080787.1 | NP_542965.1 | 3 | 633 | ||
| XM_008765012.1 | XP_008763234.1 | 3 | 1413 | |||
| XM_008765013.1 | XP_008763235.1 | 3 | 1413 | |||
| XM_008765014.1 | XP_008763236.1 | 3 | 1413 | |||
| XM_008765015.1 | XP_008763237.1 | 3 | 1255 | |||
| XM_008765016.1 | XP_008763238.1 | 3 | 496 | |||
| XM_008765017.1 | XP_008763239.1 | 3 | 1413 | |||
| XM_008765018.1 | XP_008763240.1 | 3 | 1413 | |||
| XM_008765019.1 | XP_008763241.1 | 3 | 1412 | |||
| XM_008765020.1 | XP_008763242.1 | 3 | 1249 | |||
| GenBank mRNA | FQ209812.1 | - | 3 | 437 | ||
| S49760.1 | - | 3 | 633 | |||

Dgka
Genomic Map