siRNA Details
Gene Symbol: Msmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080886.1 | NP_543162.1 | 2 | 184 | ||
GenBank mRNA | BC063155.1 | - | 2 | 171 | 6918081 | |
D50559.1 | - | 2 | 184 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Msmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080886.1 | NP_543162.1 | 2 | 184 | ||
GenBank mRNA | BC063155.1 | - | 2 | 171 | 6918081 | |
D50559.1 | - | 2 | 184 |