siRNA Details
| Gene Symbol: Txnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080887.2 | NP_543163.1 | 1, 2 | 312 | ||
| XM_006254827.2 | XP_006254889.1 | 1, 2 | 311 | |||
| GenBank mRNA | AF140358.1 | - | 1, 2 | 116 | ||
| BC098908.1 | - | 1, 2 | 312 | 7386445 | ||
| FQ213590.1 | - | 1, 2 | 283 | |||
| FQ226669.1 | - | 1, 2 | 261 | |||

Txnl1
Genomic Map