siRNA Details
Gene Symbol: C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057146.1 | NP_476487.1 | 3 | 275 | ||
GenBank mRNA | U52948.1 | - | 3 | 275 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057146.1 | NP_476487.1 | 3 | 275 | ||
GenBank mRNA | U52948.1 | - | 3 | 275 |