siRNA Details
| Gene Symbol: Oxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197332.1 | NP_001184261.1 | 3, 4 | 469 | ||
| NM_001197907.1 | NP_001184836.1 | 14, 15 | 2565 | |||
| NM_057153.3 | NP_476494.2 | 11, 12 | 2166 | |||
| GenBank mRNA | AF333986.1 | - | 3, 4 | 408 | ||
| BC081744.1 | - | 3, 4 | 451 | |||
| BC166763.1 | - | 11, 12 | 2382 | 7931847 | ||
| FM123427.1 | - | 3, 4 | 537 | |||

Oxr1
Genomic Map