siRNA Details
| Gene Symbol: Decr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D00569.1 | - | 2 | 245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Decr1| Gene Symbol: Decr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D00569.1 | - | 2 | 245 | ||