siRNA Details
| Gene Symbol: Slc8a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078620.1 | NP_511175.1 | 2 | 1006 | ||
| GenBank mRNA | U53420.1 | - | 2 | 1006 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc8a3| Gene Symbol: Slc8a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078620.1 | NP_511175.1 | 2 | 1006 | ||
| GenBank mRNA | U53420.1 | - | 2 | 1006 | ||