siRNA Details
| Gene Symbol: Maged2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271109.1 | NP_001258038.1 | 3 | 268 | ||
| NM_001271110.1 | NP_001258039.1 | 3 | 296 | |||
| NM_080479.2 | NP_536727.2 | 3 | 268 | |||
| XM_006237924.2 | XP_006237986.1 | 3 | 358 | |||
| GenBank mRNA | AJ293617.1 | - | 2 | 209 | ||
| AY724507.1 | - | 3 | 281 | |||
| BC086998.1 | - | 3 | 262 | |||
| BC107467.1 | - | 3 | 268 | 7459969 | ||
| FM059132.1 | - | 3 | 296 | |||
| FQ229095.1 | - | 3 | 281 | |||

Maged2
Genomic Map