siRNA Details
| Gene Symbol: Psmb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080767.2 | NP_542945.2 | 3 | 363 | ||
| GenBank mRNA | D10729.1 | - | 3 | 299 | ||
| FQ232382.1 | - | 3 | 308 | |||
| FQ233197.1 | - | 3 | 341 | |||
| FQ234493.1 | - | 3 | 341 | |||
| FQ235267.1 | - | 3 | 352 | |||
| KC222892.1 | - | 3 | 313 | |||
| KC222893.1 | - | 3 | 313 | |||
| KC222894.1 | - | 3 | 313 | |||
| KC222895.1 | - | 3 | 313 | |||
| KC222896.1 | - | 3 | 313 | |||

Psmb8
Genomic Map