siRNA Details
| Gene Symbol: Copb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080781.2 | NP_542959.1 | 3 | 375 | ||
| XM_008759660.1 | XP_008757882.1 | 3 | 378 | |||
| XM_008759661.1 | XP_008757883.1 | 3 | 465 | |||
| GenBank mRNA | BC061882.1 | - | 3 | 375 | 5599357 | |
| X57228.1 | - | 3 | 306 | |||

Copb1
Genomic Map