siRNA Details
| Gene Symbol: Oxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197332.1 | NP_001184261.1 | 6 | 730 | ||
| NM_001197907.1 | NP_001184836.1 | 17 | 2826 | |||
| NM_057153.3 | NP_476494.2 | 14 | 2427 | |||
| GenBank mRNA | AF333986.1 | - | 6 | 669 | ||
| BC081744.1 | - | 6 | 712 | |||
| BC166763.1 | - | 14 | 2643 | 7931847 | ||
| FQ226015.1 | - | 1 | 153 | |||

Oxr1
Genomic Map