siRNA Details
| Gene Symbol: Baiap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057196.1 | NP_476544.1 | 6 | 517 | ||
| XM_006247869.2 | XP_006247931.1 | 6 | 580 | |||
| XM_006247870.2 | XP_006247932.1 | 6 | 580 | |||
| XM_006247871.2 | XP_006247933.1 | 6 | 580 | |||
| XM_006247872.2 | XP_006247934.1 | 6 | 580 | |||
| XM_008768464.1 | XP_008766686.1 | 6 | 580 | |||
| GenBank mRNA | AY037934.1 | - | 6 | 517 | ||
| BC074009.1 | - | 6 | 551 | 7099884 | ||

Baiap2
Genomic Map