siRNA Details
| Gene Symbol: Brinp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080482.2 | NP_536730.1 | 7 | 1998 | ||
| XM_006238272.2 | XP_006238334.1 | 8 | 2416 | |||
| GenBank mRNA | AB051356.1 | - | 7 | 1999 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brinp1| Gene Symbol: Brinp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080482.2 | NP_536730.1 | 7 | 1998 | ||
| XM_006238272.2 | XP_006238334.1 | 8 | 2416 | |||
| GenBank mRNA | AB051356.1 | - | 7 | 1999 | ||