siRNA Details
| Gene Symbol: Dnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080689.4 | NP_542420.1 | 16, 17 | 1835 | ||
| GenBank mRNA | X54531.1 | - | 16, 17 | 1835 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010065, L31397, L31396, AK144142, AK220483, and others.

Dnm1
Genomic Map