siRNA Details
| Gene Symbol: Pde11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080893.1 | NP_543169.1 | 1 | 413 | ||
| GenBank mRNA | AB059362.1 | - | 1 | 413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde11a| Gene Symbol: Pde11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080893.1 | NP_543169.1 | 1 | 413 | ||
| GenBank mRNA | AB059362.1 | - | 1 | 413 | ||