siRNA Details
| Gene Symbol: Paics | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080910.3 | NP_543186.1 | 3 | 211 | ||
| XM_006250852.2 | XP_006250914.1 | 3 | 259 | |||
| XM_006250853.2 | XP_006250915.1 | 3 | 230 | |||
| XM_006250854.2 | XP_006250916.1 | 3 | 260 | |||
| GenBank mRNA | BC072508.1 | - | 3 | 201 | 7103198 | |
| BC085711.1 | - | 3 | 211 | 7192781 | ||
| D37979.1 | - | 1 | 116 | |||

Paics
Genomic Map