siRNA Details
| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130400.2 | NP_569084.1 | 1, 2 | 180 | ||
| GenBank mRNA | AF318150.1 | - | 1, 2 | 181 | ||
| BC158583.1 | - | 107 | 9022774 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhfr| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130400.2 | NP_569084.1 | 1, 2 | 180 | ||
| GenBank mRNA | AF318150.1 | - | 1, 2 | 181 | ||
| BC158583.1 | - | 107 | 9022774 | |||