siRNA Details
Gene Symbol: Ggt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130423.2 | NP_569107.2 | 2 | 290 | ||
XM_008762300.1 | XP_008760522.1 | 2 | 2915 | |||
XM_008762301.1 | XP_008760523.1 | 2 | 686 | |||
GenBank mRNA | AF244973.1 | - | 2 | 273 | ||
CB761916.1 | - | 2 | 290 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
