siRNA Details
Gene Symbol: Hrh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131909.1 | NP_571984.1 | 3 | 840 | ||
GenBank mRNA | AF358860.1 | - | 3 | 840 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
