siRNA Details
Gene Symbol: Afap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080900.1 | NP_543176.1 | 2 | 164 | ||
XM_008770245.1 | XP_008768467.1 | 3 | 323 | |||
XM_008770246.1 | XP_008768468.1 | 3 | 323 | |||
XM_008770247.1 | XP_008768469.1 | 3 | 326 | |||
GenBank mRNA | AY063759.1 | - | 2 | 164 |
