siRNA Details
| Gene Symbol: Sdf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130412.2 | NP_569096.2 | 2 | 320 | ||
| XM_006239516.2 | XP_006239578.1 | 2 | 368 | |||
| GenBank mRNA | AF405545.1 | - | 1 | 230 | ||
| BC086996.1 | - | 2 | 320 | 7192272 | ||
| FQ218523.1 | - | 2 | 345 | |||
| FQ219323.1 | - | 2 | 347 | |||
| FQ232476.1 | - | 2 | 343 | |||

Sdf4
Genomic Map