siRNA Details
| Gene Symbol: Slc36a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130415.1 | NP_569099.1 | 7 | 791 | ||
| XM_008767657.1 | XP_008765879.1 | 6 | 2890 | |||
| XM_008767659.1 | XP_008765881.1 | 8 | 1473 | |||
| GenBank mRNA | AF361239.1 | - | 7 | 791 | ||

Slc36a1
Genomic Map