siRNA Details
| Gene Symbol: Dcbld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130419.1 | NP_569103.1 | 3 | 494 | ||
| GenBank mRNA | AF387549.1 | - | 3 | 494 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcbld2| Gene Symbol: Dcbld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130419.1 | NP_569103.1 | 3 | 494 | ||
| GenBank mRNA | AF387549.1 | - | 3 | 494 | ||