siRNA Details
| Gene Symbol: Lcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130421.2 | NP_569105.1 | 3, 4 | 350 | ||
| GenBank mRNA | AB072980.1 | - | 3, 4 | 190 | ||
| BC100066.1 | - | 3, 4 | 350 | 7387128 | ||
| FQ232596.1 | - | 3, 4 | 364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lcp2| Gene Symbol: Lcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130421.2 | NP_569105.1 | 3, 4 | 350 | ||
| GenBank mRNA | AB072980.1 | - | 3, 4 | 190 | ||
| BC100066.1 | - | 3, 4 | 350 | 7387128 | ||
| FQ232596.1 | - | 3, 4 | 364 | |||