siRNA Details
| Gene Symbol: Ggt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130423.2 | NP_569107.2 | 5 | 732 | ||
| XM_008762300.1 | XP_008760522.1 | 5 | 3357 | |||
| XM_008762301.1 | XP_008760523.1 | 5 | 1128 | |||
| GenBank mRNA | AF244973.1 | - | 5 | 715 | ||

Ggt7
Genomic Map