siRNA Details
| Gene Symbol: Tmprss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130424.2 | NP_569108.2 | 5 | 477 | ||
| XM_008768547.1 | XP_008766769.1 | 5 | 593 | |||
| XM_008768548.1 | XP_008766770.1 | 5 | 524 | |||
| XM_008768549.1 | XP_008766771.1 | 5 | 706 | |||
| GenBank mRNA | AB073550.1 | - | 3 | 375 | ||
| BC061712.1 | - | 5 | 477 | 5598010 | ||

Tmprss2
Genomic Map